missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Latex Powdered Protective Gloves

Product Code. 11743017
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from high quality soft latex film, Semperguard™ Latex Powdered Protective Gloves are lightly powdered and have a microroughened outer surface.

  • Great donability, even in a humid environment
  • High-quality natural latex provides excellent comfort and fits the hand perfectly
  • Free from phthalates / softeners
  • Excellent value for money wherever powdered products are required
  • CE category III protective glove for complex risks, CE class I examination glove
  • Ideal for use with fatty foodstuff and other fatty substances
  • Tested and certified according to 1935/2004 - suitable for food contact
  • Convenient dispenser box holds 100 ambidextrous gloves (Size XL = 90 ct.)

Specifications

Certifications/Compliance CE Class I, PPE Category III
Classification Tested and certified according to 1935/2004
Coating Material Smooth inside, textured finger outside
Color Natural
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Ambidextrous
Material Rubber
Powdered Powdered
Size Small
Sterility Non-sterile
Thickness (Metric) 0.2 mm
Finger Style Textured
For Use With (Application) Suitable for food contact
Glove Cuff Style Rolled
Packaging Type Dispenser box
Palm Type Flat
Quantity 100/Pk.
Series Powdered Latex
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'