missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Style Nitrile Gloves

Product Code. 15573536
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The Black Nitrile Glove. Semperit™ Semperguard™ Style Nitrile Gloves are non-sterile, unpowdered, latex-free black nitrile gloves with excellent stretchability, a rolled cuff for easy donning and textured fingers for improved grip.

  • Black nitrile glove with excellent stretchability
  • Particularly suitable for front-end applications in the catering, beauty and body art sector
  • Thanks to dual labeling, also meets the medical requirements of the tattoo and piercing sector
  • Free from phthalates / plasticizers and allergenic latex proteins
  • Rolled cuff enables easy donning
  • Convenient dispenser box holds 100 ambidextrous gloves
  • CE category III protective glove for complex risks, CE class I examination glove
  • Tested and certified according to 1935/2004 - suitable for food contact

Specifications

Certifications/Compliance Conforms to standards EN 420, EN 374, EN455
Classification CE Category III Protective Glove for Complex Risks, CE Class I Examination Glove 
Color Black
Disposable Single-use
Exterior Finish Fingertip Textured
Grade Certified according to VO 1935/2004 - suitable for contact with food
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (Metric) 0.14 mm
For Use With (Application) Especially suitable for applications which are visible to the public
Glove Cuff Style Beaded/Rolled
Model Style Nitrile
Packaging Type Dispenser box
Quantity 100/bx
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.