Learn More
Tocris Bioscience™ Senexin A
Cdk8 inhibitor
Brand:  Tocris Bioscience™ 4875/10
239.30 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18732168
  • 303.00 € / 10mg
Explore more special offers
Description
Description
Senexin A is a cyclin-dependent kinase 8 (cdk8) inhibitor (IC50 = 280 nM). Also binds to cdk19 in an ATP-competitive manner. Inhibits p21-induced transcription. Reverses doxorubicin -induced tumor-promoting paracrine activities in vivo.
Specifications
Specifications
>98%
10 mg
310.78
1780390-76-2
Senexin A
Senexin A