Learn More
Thermo Scientific™ Sensaphone™ Telephone Dialing Systems
Remotely monitor equipment from any standard touch-tone phone with this telephone dialing system will report the status of every monitored condition.
Brand:  Thermo Scientific™ 400182
 View more versions of this product
Product Code. 12773838
  • 1407.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Alert:
Not approved for sales requiring CE mark.
Description
Description
Call the lab or office from anywhere and Thermo Scientific™ Sensaphone™ Telephone Dialing Systems will state the status of every monitored condition. These systems interface with any standard touch-tone phone to notify of alarm conditions.
  • Can be used with Model 1535 Monitor/Alarm System that features programmable audible/visual alarms and updates/displays equipment conditions on an LCD readout
  • Battery back-up is standard
  • Battery life up to 15 hr. after AC power has failed; up to 18 months with AC power on
Requires:
Six D-size alkaline batteries
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Four pre-programmed numbers
Model 1535 Monitor/Alarm System
220 V
50 Hz
Four Channels
Alarm
220 V 50 Hz