Learn More
Ansell™ HyFlex™ 48-105 Series White Polyurethane Light Weight Gloves
Designed for light applications that require good levels of comfort and abrasion resistance.
Brand: Ansell™ 48105100
27.30 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Industrial gloves
- Comfort and protection for lightweight applications
- Offers precise handling of small objects in dry or light oily environments
- Provides enhanced safety, dexterity and durability
Applications:
- Assembling, inspecting and packing small components
- Light assembly tasks
Specifications
Polyurethane | |
Nylon | |
10 | |
Knit | |
12 Pairs | |
48-105 |
Standard CATEGORY II EN 388 : 0030 | |
Lightweight Gloves | |
White | |
Nylon | |
Abrasion resistance |