missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Sensilite™ 48-135 Series White and Gray Polyurethane Light Weight Gloves

Product Code. 15260009
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15260009

Brand: Ansell™ 48135100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Meets most electrostatic discharge protection needs and provides flexibility and dexterity.

  • Special purpose gloves
  • 15-gauge nylon liner
  • Added carbon fibre offers economical product-protection solution
  • Comfort and good fit are assured through seamless construction and a wide range of sizes
  • With the palm dipped in a polyurethane coating, offers good mechanical resistance and durability
  • Fingertip-dipped version allows better fit and flexibility
  • Coated fingertip also prevents the transfer of perspiration to the electronic part

Applications:

  • Final assembly of dashboard, cables and wires
  • Final assembly of electronic components and mother boards
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 113X
Cuff Material Polyurethane
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyurethane
Product Type Lightweight Gloves
Size 10
Finger Style Coated-fingertip
Glove Cuff Style Knit
Liner Material Nylon Carbon Fiber
Lining Lined
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Good mechanical resistance
Series 48-135
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.