missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ Companion Animal Gram Positive COMPGP1F Vet AST Plate

Product Code. 18827161 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10/Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18827161

Brand: Thermo Scientific™ COMPGP1F

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Expand antimicrobial susceptibility testing of Gram positive organisms with ranges that meet CLSI Vet Companion Animal recommendations.

Perform accurate antimicrobial susceptibility testing (AST) of Gram-positive infections in companion animals with the Thermo Scientific™ Sensititre™ Companion Animal Gram Positive COMPGP1F AST Plate. Access comprehensive species-specific drug ranges which comply with the latest Veterinary CLSI recommendations to expand treatment options for serious skin infections.

Perform pradofloxacin susceptibility testing of Gram-positive organisms that cause skin infections in companion animals using the Sensititre Companion Animal Gram Positive COMPGP1F AST Plate. 

  • Robust: superior reproducibility for accurate results the first time, thereby, reducing unnecessary testing and protocols 
  • Up-to-date: includes the latest CLSI veterinary MIC ranges for companion animal Gram-positive AST to ensure relevant results based on real-time epidemiology trends
  • Easy to store: room-temperature storage eliminates inventory control concerns
  • Economic: individual packaging allows you to test one plate at a time with no waste
  • Quality assured: inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Flexible: automated or manual read options to suit your workflow

The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes. 

  • Scalable: flexible instrumentation options, designed to be modular, meet the unique needs of individual lab space, workload, and budget requirements
  • Tailor-made: design your own AST plate to meet specific formulary and surveillance requirements from a selection of more than 240 antimicrobials, including 50+ veterinary-specific agents, such as pradofloxacin.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

Specifications

0.5 McFarland Standard Demineralized water (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description Sensititre Companion Animal Gram Positive MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Gram-positive Organisms
Test Type Veterinary AST, Minimum Inhibitory Concentration (MIC)
For Use With (Application) Gram positive organisms from companion animals
Incubation Conditions 34-36°C; non-CO2
Incubation Time 18-24 hours
Isolates Per Plate 1
Quantity 10/Pk.
Reading Options Autoread or manual
Specimen Type Gram Positive Isolates Causing Skin Infections in Companion Animals
Transfer Volume 10µL
Volume Per Well 50 μL
Species Companion Animals
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.