Learn More
IKA™ Sensor Receptacle
Use to install the temperature sensors PT 100.25 and PT 100.5.
Brand:  IKA™ 0004664400
Product Code. 16228261
  • 560.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with PT 100.25 and PT 100.5 temperature sensors
  • Seal Material: FKM
Specifications
Specifications
FKM
0.166 kg
PT 100.25 and PT 100.5 Temperature Sensors