Learn More
Cytiva (formerly Pall Lab) Sentino™ Funnel Adapter
Accessory adapter for Sentino filter funnels.
Brand:  Cytiva (formerly Pall Lab) 4891
Product Code. 17124441
  • 565.00 € / Pack of 3
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For microbiology manifold
  • Lab manifold designed for flexibility and reduction in cross-contamination with interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black Fluorocarbon Elastomer O-ring
Stainless Steel (316L)
Non-sterile
3/Pk.