Learn More
Cytiva (formerly Pall Lab) Sentino™ Microbiology Pump Replacement Adapter
Pump replacement adapter for use with MicroFunnel filter funnels.
Brand:  Cytiva (formerly Pall Lab) 4872
Product Code. 17144431
  • 163.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For vacuum filtration of liquids
  • For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MicroFunnel™ Filter Funnel
Stainless Steel (316L)
Non-sterile