Learn More
BUCHI Sepacore™ Flash Systems
Brand:  BUCHI 045221
Product Code. 12724470
Description
Description
The Gradient flash system offers more flexibility than the basic flash system, yet is still very simple to use. Utilising two pumps and the programmable C-615 pump manager, the system is suitable for creating stepwise or linear gradients according to time segments. As with all Sepacore™ systems, it is compatible with virtually all mobile phases or solvent types including salt solutions and both normal and reverse phase solvents. If desired the system can easily be upgraded to include automatic peak detection and collection.
System comprises:
1 x 45230 Gradient flash system (2 x C-601 pump, C-615 pump manager, system stand, all fittings and ferrules)Please note, system supplied without separation cartridges, collection rack or tubes. Please order separately if required.
  • Simple flash system for gradient separations
  • Suitable for work with all solvent types
  • Up to 10 gradient steps per run, storage of up to 10 methods
  • Modular system, can be expanded to included detection and automatic fraction collection
  • Simple to program, reproducible results, easy to use
This robust system offers the simplicity of open column separations with the convenience of cartridges, but with the added advantages that much higher flow rates are achievable and run times are shortened dramatically. All components are chemically resistant and biocompatible. System comprises:1 x 45221 Basic flash system (C-601 pump, C-610 controller, system stand, all fittings and ferrules)1 x 45256 4 way injection/purge device1 x 44814 12mm cartridge adapter set1 x 44815 40mm cartridge adapter setPlease note, system supplied without separation cartridges, collection rack or tubes. Please order separately if required.
  • Simple flash system for isocratic separations
  • Flexible, self contained, compact and portable
  • Works with a wide range of different manufacturer's cartridge or glass column systems
  • Ideal for the replacement of open glass columns or gas pressurised systems
  • Fast, reliable, reproducible and easy to use
  • Entry level system
Specifications
Specifications
305 mm
160 mm
Ideal The Replacement of Open Glass Columns or Gas Pressurized Systems
50/60 Hz
230 V
Flash System
0 to 10 bar