Learn More
Lenz Laborglasintrumente Separating Funnels, Acc. to Squibb, Amber, with PTFE Plug, without Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 04018137
 View more versions of this product
Product Code. 11903461
  • 219.00 € / Pack of 2
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Without scale
  • With PTFE plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Specifications
100 mL
Borosilicate Glass (3.3)
NS 19/26
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Separating Funnels, Acc. to Squibb, Amber, with PTFE Plug, without Scale > 100 mL