Learn More
Lenz Laborglasintrumente Separating Funnels, Conical, Amber, without Scale, with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 04008128
 View more versions of this product
Product Code. 11963441
  • 207.00 € / Pack of 2
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Specifications
50 mL
Borosilicate Glass (3.3)