Learn More
Lenz Laborglasintrumente Separating Funnels, Conical, Amber, without Scale, with Solid Clear Glass Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 04008058
 View more versions of this product
Product Code. 11933441
  • 303.00 € / Pack of 2
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Without scale
  • With solid clear glass plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Specifications
500 mL
Borosilicate Glass (3.3)