missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephacryl™ S-100 High Resolution Media
Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules
Brand: Cytiva 17-0612-10
199.50 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Chromatography media offer good recovery and low nonspecific adsorption. Enable gel filtration at laboratory and industrial scale, and may be purchased in prepacked columns.
- For separating peptides and small proteins
- Excellent purification over a wide molecular weight range
- High reproducibility due to long-term chemical and physical stability
- High flow rates and recoveries
- Well-suited to industrial-scale use
- Storage conditions: 20% ethanol; 4° to 30°C
- Available in a variety of selectivities and volumes
Chemical Stability (stable in all common buffers):
- 1M acetic acid, 8M urea
- 6M guanidine-HCl, 1% SDS
- 2M NaCl, 24% ethanol
- 30% propanol, 30% acetonitrile
- 0.5M NaOH (for cleaning)
- 3 to 11 (long-term and working)
- 2 to 13 (short-term)
- Negligible volume variation due to changes in pH or ionic strength
- Autoclavability: At 121°C, pH 7 for 30 minutes (gel only)
- Antimicrobial agent: 20% ethanol
Specifications
150mL | |
25 to 75 μm | |
Sephacryl HR Media | |
40 to 140cm | |
5.9 in. | |
100kPa | |
Allyl Dextran and N, N'-methylenebisacrylamide | |
4° to 30°C |
>125 cm/hr. | |
19 to 230L | |
Solid Spherical Bead | |
Allow fast and reproducible purification of peptides and small proteins by gel filtration at laboratory and industrial scale | |
15cm | |
3 to 11 | |
Peptides and small proteins |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sephacryl S-100 High Resolution, 150 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.