missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ S-500 High Resolution Media

Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules

Brand:  Cytiva 17-0613-10

197.00 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 11560794

  • 216.00 € / 150mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Chromatography media offer good recovery and low nonspecific adsorption. Enable gel filtration at laboratory and industrial scale, and may be purchased in prepacked columns.

  • For separating polysaccharides, macromolecules with extended structures, and small plasmids
  • Excellent purification over a wide molecular weight range
  • High reproducibility due to long-term chemical and physical stability
  • High flow rates and recoveries
  • Well-suited to industrial-scale use
  • Storage conditions: 20% ethanol; 4° to 30°C
  • Available in a variety of selectivities and volumes

Chemical Stability (stable in all common buffers):

  • 1M acetic acid, 8M urea
  • 6M guanidine-HCl, 1% SDS
  • 2M NaCl, 24% ethanol
  • 30% propanol, 30% acetonitrile
  • 0.5M NaOH (for cleaning)
pH Stability
  • 3 to 11 (long-term and working)
  • 2 to 13 (short-term)
Physical Stability:
  • Negligible volume variation due to changes in pH or ionic strength
  • Autoclavability: At 121°C, pH 7 for 30 minutes (gel only)
  • Antimicrobial agent: 20% ethanol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

150 mL
25 to 75 μm
Sephacryl HR Media
40 to 140cm
5.9 in.
100kPa
Allyl Dextran and N, N'-methylenebisacrylamide
4° to 30°C
>125 cm/hr.
19 to 230L
Solid Spherical Bead
Allow fast and reproducible purification of proteins, polysaccharides and other macromolecules by gel filtration at laborato ry and industrial scale
15cm
3 to 11
Polysaccharides, macromolecules and small plasmids
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sephacryl S-500 High Resolution

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.