missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G-100 Media
Commonly used for desalting and buffer exchange of very large biomolecules
Brand: Cytiva 17-0060-01
630.80 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in two particle size ranges
Fine particle size
- Particle size, dry: 40 to 120μm
- Fractionation range, globular proteins: 4000 to 150,000
- Particle size, dry: 10 to 40μm
- Fractionation range, globular proteins: 4000 to 100,000
Specifications
100 g | |
100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form) | |
Sephadex G-100 Media | |
Useful for desalting and Buffer exchange of very large biomolecules | |
2 to 10 | |
Large biomolecules |
4.2 mL/min. | |
15 to 20mL/g in Distilled Water | |
Solid Bead | |
0.096 bar | |
Cross-linked Dextran | |
4° to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction