missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G10 Media
Gel filtration medium for desalting and buffer exchange of peptides and small biomolecules
Brand: Cytiva 17-0010-01
315.28 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Beads behave as rigid spheres obeying Darcy's Law for purposes of determining maximum flowrate and maximum operating pressure
- Working pH range 2 to 13
- Particle size range 55 to 165μm
- Bed volume in distilled water 2-3mL/g
- Exclusionation limit: 7 x 102 dextrans, 7 x 102 globular proteins
Specifications
100 g | |
55 to 165 μm in wet form, 40 to 120 μm in dry form | |
Sephadex G10 Media | |
2.6cm | |
11.81 in. | |
24-49 cm/h, pressure drop cm H2O/bed height = 2, bed height 30 cm, 2.6 cm i.d | |
Cross-linked Dextran | |
4 to 30°C |
< Darcy´s Law | |
2 to 3mL/g in Distilled Water | |
Solid Bead | |
Desalting and Buffer exchange of Peptides and small biomolecules >700 M.W. | |
30cm | |
2 to 13 | |
Peptides, Small Biomolecules |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction