missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G50 Fine Media

Suitable for gel filtration

445.00 € - 1918.00 €

Specifications

Flow Rate < Darcy´s Law
Particle Size 34 to 208 μm in wet form, 20 to 80 μm in dry form
Bed Volume 9 to 11mg/g
Product Type Sephadex G50 Media
Format Solid Bead
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11534875
View Documents
Cytiva
17-0042-01
100 g
445.00 €
100g
In Stock
to see stock.
Add to basket
 
11544875
View Documents
Cytiva
17-0042-02
500 g
1918.00 €
500g
In Stock
to see stock.
Add to basket
 
Description

Description

  • Particle size range 40 to 160μm

Specifications

Specifications

< Darcy´s Law
9 to 11mg/g
Solid Bead
For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids
10cm
2 to 10
DNA
34 to 208 μm in wet form, 20 to 80 μm in dry form
Sephadex G50 Media
5cm
3.93 in.
min 150 cm/h, bed height 10 cm, column 5 cm i.d.
Cross-linked Dextran
4 to 30°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.