missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G50 Medium Media

Suitable for gel filtration

Brand:  Cytiva 17-0043-01

365.75 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 11554875

  • 396.00 € / 100g
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Particle size range of wet beads: 100 to 300μm
  • Fractionation range (Mr), globular proteins: 1500 to 30,000

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100g
100 to 300 μm in wet form, 50 to 150 μm in dry form
Sephadex G50 Media
5cm
3.93 in.
min 150 cm/h, bed height 10 cm, column 5 cm i.d.
Cross-linked Dextran
4 to 30°C
< Darcy´s Law
9 to 11mg/g
Solid Bead
For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids
10cm
2 to 10
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers