missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G75 Media
Suitable for gel filtration
Brand: Cytiva 17-0050-01
416.10 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- For purposes of determining maximum flowrate, beads behave as rigid spheres according to Darcy's Law
- Maximum operating pressure 0.15 bar
- Working pH range 2 to 10
- Particle size range of wet beads 90 to 280μm
- Bed volume in distilled water 12 to 15mL/g
- Fractionation ranges: 1 x 103 to 5 x 104 for dextrans, 3 x 103 to 8 x 104 for globular proteins
Specifications
100 g | |
90 to 280 μm in wet form, 40 to 120 μm in dry form | |
Sephadex G75 Media | |
5cm | |
3.93 in. | |
min 90 cm/h, bed height 10 cm, column 5 cm i.d. | |
Cross-linked Dextran | |
4 to 30°C |
< Darcy´s Law | |
12 to 15mg/g | |
Solid Bead | |
Desalting and Buffer exchange of large biomolecules >80000 M.W. | |
10cm | |
2 to 10 | |
DNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction