missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G75 Superfine Media
Classic gel filtration medium
Brand: Cytiva 17-0051-01
This item is not returnable.
View return policy
Description
- Fractionation range, globular proteins: 3000 to 70,000
- Particle size, wet: 20 to 155μm
- Working pH range: 2-10

Specifications
100 g | |
22 to 143 μm in wet form, 10 to 40 μm in dry form | |
Sephadex G75 Media | |
Desalting and Buffer exchange of large biomolecules, for specific cases requiring very high resolution | |
2 to 10 | |
DNA |
1.5 mL/min. | |
12 to 15mg/g | |
Solid Bead | |
Care must be taken to avoid compressing the gel. Do not increase the pressure beyond 0.16 bar. | |
Cross-linked Dextran | |
4° to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction