missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ Ion Exchange Media

Separate molecules over broad molecular weight and pH ranges

Brand:  Cytiva 17-0210-02

 View more versions of this product

Product Code. 11524895

  • 2166.00 € / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.

Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use

DEAE Sephadex

  • Weak and strong anion exchanger
  • pH range 2-9

QAE Sephadex
  • Strong anion exchanger
  • pH range: 2-12
CM Sephadex
  • Weak cation exchanger
  • pH range: 6-10
SP Sephadex
  • Strong cation exchanger
  • pH range: 4-13

TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 g
Column or Batch Techniques
Solid Spherical Bead
2 to 5.9 in.
Carboxymethyl
6 to 10
Up to 30,000
40 to 120 μm in dry form
Sephadex Ion Exchange Media
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
5 to 15cm
CM-Sephadex C-25
Cross-linked Dextran
4° to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers