missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ Ion Exchange Media
Separate molecules over broad molecular weight and pH ranges
Brand: Cytiva
This item is not returnable.
View return policy
Description
Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.
Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use
DEAE Sephadex
- Weak and strong anion exchanger
- pH range 2-9
QAE Sephadex
- Strong anion exchanger
- pH range: 2-12
- Weak cation exchanger
- pH range: 6-10
- Strong cation exchanger
- pH range: 4-13
Specifications
100 g | |
Column or Batch Techniques | |
Solid Spherical Bead | |
2 to 5.9 in. | |
Sulphopropyl | |
4 to 13 | |
Up to 30,000 |
>40 μm | |
Sephadex Ion Exchange Media | |
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques | |
5 to 15cm | |
SP-Sephadex C-25 | |
Cross-linked Dextran | |
4° to 30°C |