missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sepharose Agarose Gel Filtration Media
Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.
Brand: Cytiva 17-0120-01
This item is not returnable.
View return policy
Description
European Pharmacopoeia Solutions
- Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
- All are manufactured and tested in compliance with the Ph. Eur.
- Supplied with a Certificates of Analysis
- Lot No. and expiry date are stated on all product labels
Order Info
Supplied in 1L plastic containers
Specifications
1 L | |
45 to 165 μm | |
Solid Bead | |
Allow characterization or cleaning up of samples containing components of diverse molecular weight | |
10cm | |
4 to 9 | |
4 to 30°C, 20% Ethanol |
<11.5 cm/hr. | |
Sepharose Agarose Gel Filtration Media | |
5cm | |
3.94 in. | |
70-140 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, column 5 cm i.d. | |
4% Agarose |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- sepharose 4B
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction