Learn More
Cytiva Sepharose™ Fast Flow Cation Exchange Media
SP Sepharose Fast Flow is a sulphopropyl (SP) strong cation exchange chromatography resin for fast protein purification. CM Sepharose Fast Flow is a weak cation exchanger based on the well established Sepharose Fast Flow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.
Brand: Cytiva 17-0719-10
View more versions of this product
Description
Used for preparative protein separations in both research and industrial applications. Cytiva CM Sepharose™ Fast Flow Cation Exchange Media is composed of crosslinked 6% agarose beads, with carboxymethyl (CM) weak cation exchange groups.
- Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
- Composed of crosslinked 6% agarose beads
- Reproducible
- Part of Sepharose Fast Flow ion exchange platform
- High chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols
- Available in range of pre-packed process development tools
SP Sepharose:
- Ligand: Sulphopropyl
- Ion Exchanger Type: Strong cation exchanger
- Ionic Capacity: 0.18 to 0.25mmol H+/mL medium
- Binding capacity: 70mg ribonuclease A/mL medium
- Ligand: Carboxymethyl
- Ion Exchanger Type: Weak cation exchanger
- Ionic Capacity: 0.09 to 0.13mmol Hl+/mL medium
- Binding capacity: 50mg ribonuclease A/mL medium
Used extensively for capture and intermediate purification of wide range of approved biopharmaceuticals
Specifications
50 mg ribonuclease A/mL mediμm | |
45 to 165 μm | |
25 mL |
CM Sepharose Fast Flow Media | |
4 to 13 |
Safety and Handling
- CM Sepharose Fast Flow, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.