Learn More
Thermo Scientific™ Septa
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Brand:  Thermo Scientific™ 313G3211
 View more versions of this product
Product Code. 11998409
  • 83.66 € / Pack of 50
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Low Coring Septum
Trace and Focus GC Instruments
17 mm
TR-Green; 17 mm dia.; Max. Temp. 350°C
Silicone
50 (Glass Jar)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.