Learn More
Fisherbrand™ Septum for 10-425 screw thread cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
43.90 € - 490.00 €
Products 5
Product Code Brand Color Material Price Quantity & Availability  
Product Code Brand Color Material Price Quantity & Availability  
11350850
View Documents
Fisherbrand™
09 02 1260
Red, White PTFE, Silicone
262.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
11834820
View Documents
Fisherbrand™
09 02 1261
Red, White PTFE, Silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11824820
View Documents
Fisherbrand™
09 02 1259
Red-Orange, Transparent Natural Rubber, TEF This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10422614
View Documents
Fisherbrand™
10 02 0646
White PTFE Virginal This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11844820
View Documents
Fisherbrand™
09 02 1327
Blue, White PTFE, Silicone
490.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
Description
Description
  • Septa