missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 8-425 screw thread cap

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™ 08 02 1966

79.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16160961

  • 84.45 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 8mm Septa
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Septa
8 mm
1000
Beige, Red-Orange
PTFE
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers