missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequence Detection Systems Spectral Calibration Kit, 384-well

Easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 384-well block of the 7900HT Sequence Detection System quickly and easily.

Brand:  Applied Biosystems™ 4323977

488.70 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10239674

  • 1086.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Applied Biosystems™ easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™,ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™ dyes on the 7900HT Sequence Detection System. The Sequence Detection Systems Spectral 384-well Calibration kit includes two optical 384-well reaction plates: one preloaded and sealed background plate, and one preloaded and sealed spectral calibration plate with eight separate dye standards. Spend Your Time on Research - Not Instrument Calibration This kit's easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 384-well block of the 7900HT Sequence Detection System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

  • 96-well or 384-well optical reaction plates preloaded with dyes (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™) are provided for quick and easy calibration of the ABI PRISM™ 7900HT Sequence Detection System
  • Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
  •  
    For Research Use Only. Not for use in diagnostic procedures.

    Order Info

    Expiration date: 12 months from the date of delivery.

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Includes one Background and one Spectral Calibration plate with eight dye standards plates (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™).

    Store at -15 to 30°C. Protect from light.

    Expiration date: 12 months from the date of delivery
    AB 7900 HT 384-well System
    Spectral Calibration Kit
    Dry Ice
    2 x 384-well Plates
    FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
    Fluorescence
    Real Time PCR (qPCR)
    384-well Plate
    Spectral Calibration
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.