missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophilic

Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

Brand:  Cytiva 24152105050250

370.97 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 11588712

  • 417.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophilic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5% solids (50mg/mL)
DNA Sample Prep, Protein Purification
1 μm
2°C to 8°C
15 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers