missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads

Suited for capturing or isolating mRNA from a variety of sources. Due to their cauliflower like structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.

Brand:  Cytiva 38152103010150

508.45 EUR valid until 2024-12-31
Use promo code "21948" to get your promotional price.


 View more versions of this product

Product Code. 12628335

  • 575.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
  • High binding capacity
  • 1.0μm diameter
  • High oligo dT content for mRNA binding
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1% solids (10mg/mL)
mRNA Purification
1 μm
2°C to 8°C
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers