Learn More
Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
Brand:  Cytiva 19152104011150
 View more versions of this product
Product Code. 11854962
  • 205.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics
  • Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Amine
    Magnetite
    1 μm
    10 mg/mL
    1 mL