Learn More
Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads
Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).
Brand:  Cytiva 17152104010150
500.82 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 15732442
  • 577.00 € / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1% solids (10mg/mL)
Affinity Purification
1 μm
2°C to 8°C
5 mL