Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, low biotin binding capacity
Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.
Brand:  Cytiva 30152103010350
 View more versions of this product
Product Code. 11855012
  • 3157.00 € / Each
Explore more special offers
Description
Description
Biotin Binding Capacity of 2500 to 3500pmol/mg beads
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1% solids (10mg/mL)
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
1 μm
2°C to 8°C
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, low biotin binding capacity > 100mL