missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Xtracta Virus/Pathogen Kit

Sera-Xtracta Virus/Pathogen Kit for high-throughput total nucleic acid (DNA/RNA) isolation from bacteria and viruses including Adenovirus (Type 14), Influenza A (H3N2) and COVID-19.

Brand:  Cytiva 29514201

2272.40 EUR valid until 2024-12-31
Use promo code "21969" to get your promotional price.



Product Code. 16690284

  • 2464.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Scalability: Up to 400 μL of sample
  • Recovery Specifications: Reproducible detection of low viral titer from 10 pfu/mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

All reagents can be stored at ambient temperature (15 to 25°C). For Proteinase K, once reconstituted, store at 2 to 8°C
630 mL Binding/Lysis Reagent; 1000 mL Wash Buffer ; 10.4 mL SeraSil-Mag, 400 beads; 10.4 mL SeraSil-Mag, 700 beads; 2 x 110 mg Proteinase K (lyophilized)
Can be completed in less than 30 minutes
Nucleic acid extraction
1000 Extractions
Liquid, Magnetic Bead Based
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SeraXtracta Virus/Pathogen Kit (1000 extractions)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : proteases, salts ofthiocyanic acid, hydrogen chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers