missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
Brand: BINDER™ 9640-0013
11977.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Optimized double-pan humidification system with condensation protection
- Tried-and-tested anti-contamination concept with 180°C hot air sterilization
- Hot-air sterilizable CO2 sensor with single-beam infrared technology
- Seamless inner chamber made of stainless steel with flanges as shelf support system
- Fanless interior with Venturi CO2 gas mixing nozzle
- Intuitive touchscreen controller
- Internal data recording, can be read out in open format via USB interface
- Fault diagnosis system with visual and acoustic alarm
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Fail-safe CO2 system for protecting the pH of cell cultures
Specifications
9.4 cu. ft. | |
10 kg | |
267 L | |
0 to 20% | |
Fanless interior | |
None | |
Stainless Steel | |
3 | |
None | |
Ambient +4° to 60°C | |
TFT touch | |
IR sensor | |
620 x 575 x 750 mm | |
125 kg | |
1.5 kW | |
50/60 Hz |
22 lb. | |
CB260-230 V, Standard, Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor, 267 L, 230 V 1 ∼ 50/60 Hz | |
Stainless Steel | |
IR Sensor | |
Natural | |
No Lighting | |
1 | |
1 to 21% | |
Nema 5-20P | |
CO2 Incubator | |
CB260-230 V | |
IR sensor | |
745 x 885 x 1020 mm | |
230 V | |
6.5 A |