missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
GREENER_CHOICE

The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.

Brand:  BINDER™ 9640-0006

 View more versions of this product

Product Code. 17393693

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Optimized double-pan humidification system with condensation protection
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data recording, can be read out in open format via USB interface
  • Fault diagnosis system with visual and acoustic alarm
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures
Specifications

Specifications

1.9 cu. ft.
10 kg
53 L
0 to 20%
Fanless interior
None
Stainless Steel
2
Yes
90 to 95%
CO2 Incubator
CB056-230 V-O
IR sensor
528 x 645 x 674 mm
230 V
3.5 A
22 lb.
CB056-230 V-O, with O2 control 0.2-20%, Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor, 53 L, 230 V 1 ∼ 50/60 Hz
Stainless Steel
IR Sensor
Natural
No Lighting
1
1 to 21%
Nema 5-20P
Ambient +6° to 60°C
TFT touch
IR sensor
400 x 330 x 400 mm
53 kg
0.8 kW
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.