missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
GREENER_CHOICE

Product Code. 17739794 Shop All BINDER Products
Click to view available options
Capacity (Metric):
53 L
170 L
267 L
Model:
CB056-230 V
CB056-230 V-O
CB170-230 V
CB170-230 V-O
CB170-230V-D
CB260-230 V
CB260-230 V-O
Oxygen Control:
None
Yes
Förpackningsstorlek
Each
Denne vare kan ikke returneres. Se returpolitik
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Produktkode 17739794

missing translation for 'mfr': BINDER™ 96400027

Kun null tilbage
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.

  • Optimized double-pan humidification system with condensation protection
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data recording, can be read out in open format via USB interface
  • Fault diagnosis system with visual and acoustic alarm
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures

Tekniske data

Capacity (English) 6 cu. ft.
Capacity (English) Shelf 22 lb.
Capacity (Metric) Shelf 10 kg
Description CB170-230V-D, with Access port 30 mm left, Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor, 170 L, 230 V 1 ∼ 50/60 Hz
Capacity (Metric) 170 L
Chamber Material Stainless Steel
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Convection Type Natural, APT.Line Technolgoy
Holds None
Lighting No Lighting
Material Stainless Steel (18/8)
No. of Places 1
No. of Shelves 3
O2 Concentration Range 1 to 21%
Oxygen Control None
Plug Type Nema 5-20P
Relative Humidity 90 to 95%
Temperature Range (Metric) Ambient +4° to 60°C
Type CO2 Incubator
Display TFT touch
Model CB170-230V-D
O2 Sensor Technology IR sensor
Sensor Types IR sensor
Dimensions (D x W x H) Interior 560 x 505 x 600 mm
Dimensions (L x W x H) 685 x 815 x 870 mm
Weight (Metric) 98 kg
Voltage 200/230 V
Wattage 1.4 kW
Amperage 5,7 A
Frequency 50/60 Hz
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.