missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series VDL Vacuum Drying Chamber for Flammable Solvents

 Binder&trade Model VDL Vacuum Drying Chamber for Flammable Solvents for fast and gentle drying of flammable organic solvents.

Specifications

Electrical Requirements 230 V, 1 phase, 50/60 Hz, 0.8 kw, 10 A
Type Drying and Heating Chamber
View More Specs

Description

Description

  • Fast and gentle drying
  • Direct heat transfer through large thermal conducting plates
  • Safe work with one-of-a-kind safety concept
  • Coordinated, modular system with vacuum pump and vacuum module
  • Temperature range: room temperature plus 15 °C to 200 °C
  • ATEX compliance chamber's interior: Ex II -/3G c IIB T3-T1 Gc X
  • APT.line™ preheating chamber technology
  • Controller with time-segment and real-time programming
  • 2 aluminum expansion racks
  • Inert gas connection
  • Shatterproof, spring-mounted safety glass panel
  • Pressurized instrument panel with compressed air connection
  • Pressure control device for heating activated < 125 mbar
  • Flame protection gasket
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Computer interface: RS 422
 
Specifications

Specifications

230 V, 1 phase, 50/60 Hz, 0.8 kw, 10 A
Drying and Heating Chamber
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.