Learn More
Fisher BioReagents™ Serine Inhibitor Cocktail I For Broad Range Serine Protease
Brand:  Fisher BioReagents™ 12861650
Product Code. 12861650
  • 70.65 € / Each
Explore more special offers
Description
Description
Vial makes 1mL 100x, contains AEBSF, Aprotinin, Elastatinal, GGACK store in freezer at -20'C Fisher Bioreagents
Specifications
Specifications
100X
1 mL