Learn More
TPP Techno Plastic Products™ Serological Pipets
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
Description
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
- Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
- Smooth interiors, prevent cell hang-up or clumping inside the pipette
- Excellent optical transparency
- Color coded cotton plugs and dispenser boxes for easy identification
- Convenient, stackable TPP dispenser box
- Single wrapped in lint-free paper/plastic blister
- Slender and short form enables ergonomic working conditions in the sterile hood
- Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
- Highly drop free pipet tip design
- Optimized form of the mouthpiece preserves the rubber of the pipet aid
- Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Specifications
Polystyrene | |
Yes |