missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12010-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.Specifications
SERPINA6 | |
Polyclonal | |
Unconjugated | |
SERPINA6 | |
CBG, Serpin A6, SERPINA6, Transcortin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
866 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08185 | |
SERPINA6 | |
SERPINA6 Fusion Protein Ag2623 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction