missing translation for 'onlineSavingsMsg'
Learn More
Gravis Serum Flasks Product Code.: 11783813
Add to basket

Gravis Serum Flasks

Product Code. 11783813
8 mL
Click to view available options
Capacity (Metric):
8 mL
15 mL
24 mL
This item is not returnable. View return policy

Product Code. 11783813

Brand: Gravis F018094672

Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Capacity (Metric) 8 mL
Diameter (Metric) Outer Neck 20 mm
Material Glass
No. per Pack 224Pack
Quantity 224Pack
Format Flask
Neck Finish 20mm
Product Type Serum Flask
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gravis Serum Flasks >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.