Learn More
Interscience™ Set of 5 Teflon™ Nozzles
Set of 5 nozzles and caps for dilutor tubings.
Brand:  Interscience™ 513024
Product Code. 15530703
  • 184.00 € / Each
Explore more special offers
Description
Description
  • For tubings with 4.8 to 8 mm inner diameter
Specifications
Specifications
Tubing Nozzle
Applying Nozzles
5 Nozzles/Set
Autoclavable
DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro