missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ SevenGo Duo™ Pro pH/Ion/Conductivity Multi Parameter Meters
Portable instruments for pH, conducitivity, ion and dissolved oxygen determination
Brand: Mettler Toledo™ 51302623
This item is not returnable.
View return policy
Description
The instruments are suited for mobile measurements both in the field and in the production environment. The SG23 SevenGo Duo™ pH/mV/conductivity meter is best suited for parallel routine measurements in dual-channel mode. In addition, it also offers Intelligent Sensor Management (ISM™). The SG78 and SG68 SevenGo Duo Pro™ instruments allow simultaneous determination and display of two electrochemical parameters with highest possible accuracy. They have the same comprehensive GLP functions as their SevenGo Pro™ counterparts. In addition, the SevenGo Duo pro™ instruments have extra security functions, such as Intelligent Sensor Management (ISM ™), PIN-protected device logon and PIN-protected system setting.- Portable instruments for pH, conductivity, ion and dissolved oxygen determination
- Robust, watertight single- and dual-channel instruments for use in demanding conditions
- Unique ergonomics and extremely simple operation
Specifications
± 0.5% | |
5 | |
0.01 μS/cm to 1000 mS/cm | |
0.01 to 100.0 Mohm/4cm | |
0.1 mg/L to 600 g/L | |
Suited for mobile measurements both in the field and in the production environment | |
SG78-FK5 | |
-1999 to 1999 mV |
1.5 V | |
IP 67 | |
-2 to 20 | |
0 to 80 psu | |
-5°C to +130°C (pH ATC), -30°C to +130°C (MTC) | |
Field Kit with InLab Expert Pro-ISM, InLab738-ISM (both IP67, 5m cable) and uGo | |
Field Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction