missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17926-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two pseudogenes, one on chromosome 15 and the other on chromosome 21, have been found for this gene.Specifications
SFRS9 | |
Polyclonal | |
Unconjugated | |
SRSF9 | |
SFRS9, SRp30c, SRSF9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108014, 288701, 8683 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13242, Q5PPI1, Q9D0B0 | |
Srsf9 | |
SFRS9 Fusion Protein Ag12347 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |