missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SFX-Insect™ Cell Culture Media
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
Brand: Cytiva SH30350.03
261.59 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Protein free media
Specifications
cGMP | |
Powder | |
Non-sterile | |
Glutamine, Poloxamer 188 | |
10 L |
2° to 8°C, away from light and moisture | |
Insect Cell Medium | |
Non-sterile | |
No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES |