missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BP1/BP1 PONA Column

SGE GC column BP1 PONA is an industry standard non polar phase suitable for all routine analysis

713.00 € - 1345.00 €

Specifications

Stationary Phase 100% Dimethyl Polysiloxane
Polarity Non polar
View More Specs

Products 4
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
15288633
View Documents
Trajan™
054082
50 m 0.32 mm 5 μm
1345.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
10675631
View Documents
Trajan™
054041
25 m 0.22 mm 0.1 μm
713.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
15208633
View Documents
Trajan™
054050
30 m 0.22 mm 0.25 μm
713.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
12985531
View Documents
Trajan™
054072
50 m 0.32 mm 1 μm
1345.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
Description

Description

  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications

Specifications

100% Dimethyl Polysiloxane
Non polar
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.