missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Diamond Thermo RSH 10 to 50μL GC Autosampler Syringes

Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.

Brand:  Trajan™ 0355620

 View more versions of this product

Product Code. 13147946

  • 115.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications

Specifications

23 G
0.24 mm
0.63 mm
Ultra Smooth Hydrophobic Coating
GC Autosampler Syringe
10 to 50 μL
Cone
0.009 in.
0.024 in.
Gas chromatography
2.24 in.
Ea.
57 mm
Diamond Thermo RSH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers